Human recombinant PSA (from cells)
Fournisseur: ProSci Inc.
92-336
PRSI92-336EA 884 EUR InStock
PRSI92-336
Human recombinant PSA (from cells)
Protéines et Peptides
KLK3, also known as APS, is short for Prostate-specific antigen. It is a 261 aa. protein which belongs to the peptidase S1 family and Kallikrein subfamily. This protein has 5 isforms produced by alternative splicing. It is a secreted protein and can forms a heterodimer with SERPINA5 which can inhibit its activity. KLK3 is also strongly inhibited by Zn²⁺, 100 times more abundant in semen than in serum. This inhibition is relieved by exposure to semenogelins, which are avid zinc binders. KLK3 can hydrolyses semenogelin-1 thus leading to the liquefaction of the seminal coagulum.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité