Human recombinant NPDC1 (from cells)
Fournisseur: ProSci Inc.
92-339
PRSI92-339EA 588 EUR InStock
PRSI92-339
Human recombinant NPDC1 (from cells)
Protéines et Peptides
Neural proliferation differentiation and control protein 1(NPDC1) is a protein that in humans is encoded by the NPDC1 gene. It is a single-pass membrane protein and belongs to the NPDC1/cab-1 family. The protein strongly expressed in adult brain and especially in hippocampus, frontal lobe and temporal lobe. The protein suppresses oncogenic transformation in neural and non-neural cells and down-regulates neural cell proliferation and it might be involved in transcriptional regulation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité