Mouse recombinant placenta growth factor
Fournisseur: ProSci Inc.
92-342
PRSI92-342EA 947 EUR InStock
PRSI92-342
Mouse recombinant placenta growth factor
Protéines et Peptides
Placental growth factor is a protein that in humans is encoded by the PGF gene. It is a secreted protein and belongs to the PDGF/VEGF growth factor family. The protein is a member of the VEGF (vascular endothelial growth factor) sub-family-a key molecule in angiogenesis and vasculogenesis, in particular during embryogenesis. The main source of PGF during pregnancy is the placental trophoblast. PGF is also expressed in many other tissues, including the villous trophoblast.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité