Mouse recombinant CX3CL1
Fournisseur: ProSci Inc.
92-343
PRSI92-343EA 654 EUR InStock
PRSI92-343
Mouse recombinant CX3CL1
Protéines et Peptides
Fractalkine(CX3CL1) is a single-pass type I membrane protein and belongs to the intercrine delta family. It consists of an extracellular NH2-terminal domain, a mucin-like stalk, a transmembrane alpha helix, and a short cytoplasmic tail. CX3CL1 exists in two forms: as a membrane-anchored or as a shed 80-95K glycoprotein. Soluble CX3CL1 is generated by limited proteolysis on the cell surface, and a disintegrin and metallopeptidase 10 (ADAM10) and ADAM17/tumour necrosis factor- alpha-converting enzyme (ADAM17/TACE) participate in this shedding. It has been suggested that ADAM10 acts in the constitutive shedding, and ADAM17 acts in response to cell activation. The protein may play a role in regulating leukocyte adhesion and migration processes at the endothelium.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité