Human recombinant TNF receptor II (from cells)
Fournisseur: ProSci Inc.
92-344
PRSI92-344EA 654 EUR InStock
PRSI92-344
Human recombinant TNF receptor II (from cells)
Protéines et Peptides
tumour necrosis factor receptor superfamily member 1B is a 461 amino acids protein that belongs to the TNFR (tumour necrosis factor receptor) superfamily Characterised by cysteine-rich extracellular domains. It contains 4 TNFR-Cys repeats. TNFRII is expressed in fetal brain. TNFRII is strongly expressed at the cartilage-pannus junction, and plays a major role in a subset of families with multiple cases of rheumatoid arthritis (RA). This receptor mediates most of the metabolic effects of TNF-alpha. Isoform 2 blocks TNF-alpha-induced apoptosis, which suggests that it regulates TNF-alpha function by antagonizing its biological activity.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité