Human recombinant PFKFB1 (from cells)
Fournisseur: ProSci Inc.
92-350
PRSI92-350EA 884 EUR InStock
PRSI92-350
Human recombinant PFKFB1 (from cells)
Protéines et Peptides
6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1 is an enzyme that in humans is encoded by the PFKFB1 gene. The enzyme forms a homodimer that catalyses both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains. It belongs to the phosphoglycerate mutase family. Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Consequently, regulating fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité