Human recombinant malate dehydrogenase (MDH) (from cells)
Fournisseur: ProSci Inc.
92-353
PRSI92-353EA 789 EUR InStock
PRSI92-353
Human recombinant malate dehydrogenase (MDH) (from cells)
Enzymes
Malate dehydrogenase, mitochondrial is a 338 amino acids protein that belongs to the LDH/MDH superfamily. MDH type 1 family. MDH2 catalyses the reversible oxidation of malate to oxaloacetate, utilising the NAD/NADH cofactor system in the citric acid cycle. MDH2 is localised to the mitochondria and takes part in the malate-aspartate shuttle that functions in the metabolic coordination between cytosol and mitochondria. MDH2 is highly expressed in the adrenal system, small intestine, heart and pancreas.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité