Human recombinant NGF beta (from cells)
Fournisseur: ProSci Inc.
92-356
PRSI92-356EA 343 EUR InStock
PRSI92-356
Human recombinant NGF beta (from cells)
Protéines et Peptides
Human beta -Nerve Growth Factor ( beta -NGF) was initially isolated in the mouse submandibular gland. It is composed of three non-covalently linked subunits alpha, beta , and gamma ; it exhibits all the biological activities ascribed to NGF. It is structurally related to BDNF, NT-3 and NT-4 and belongs to the cysteine-knot family of growth factors that assume stable dimeric structures. beta -NGF is a neurotrophic factor that signals through its receptor beta -NGF, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. beta -NGF also acts as a growth and differentiation factor for B lymphocytes and enhances B-cell survival. These results suggest that beta -NGF is a pleiotropic cytokine, which in addition to its neurotropic activities may have an important role in the regulation of the immune system. Human beta -NGF shares 90% sequence similarity with mouse protein and shows cross-species reactivity.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité