Human recombinant phosphoglycerate kinase 1 (from cells)
Fournisseur: ProSci Inc.
92-357
PRSI92-357EA 884 EUR InStock
PRSI92-357
Human recombinant phosphoglycerate kinase 1 (from cells)
Enzymes
PGK1 is called phosphoglycerate kinase that involved in a critical energy-producing process known as glycolysis. Phosphoglycerate kinase helps carry out a chemical reaction that converts a molecule called 1,3-diphosphoglycerate, which is produced during the breakdown of glucose, to another molecule called 3-phosphoglycerate during glycolysis. PGK1 The encoded protein may also act as a cofactor for polymerase alpha.. The protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité