Human recombinant EGFR (from cells)
Fournisseur: ProSci Inc.
92-359
PRSI92-359EA 588 EUR InStock
PRSI92-359
Human recombinant EGFR (from cells)
Protéines et Peptides
EGFR is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerisation and tyrosine autophosphorylation and leads to cell proliferation. Activates at least 4 major downstream signalling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STATs modules. May also activate the NF-kappa-B signalling cascade. Also directly phosphorylates other proteins like RGS16, activating its GTPase activity and probably couplling the EGF receptor signalling to the G protein-coupled receptor signalling.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité