Human recombinant IFN gamma (from cells)
Fournisseur: ProSci Inc.
92-362
PRSI92-362EA 409 EUR InStock
PRSI92-362
Human recombinant IFN gamma (from cells)
Protéines et Peptides
IFN gamma is the major interferon produced by mitogenically or antigenically stimulated lymphocytes. It is structurally different from type I interferon and its major activity is immunoregulation. It has been implicated in the expression of class II histocompatibility antigens in cells that do not normally produce them, leading to autoimmune disease. Interferon gamma is produced mainly byT-cells and natural killer cells activated by antigens, mitogens, or alloantigens. It is produced by lymphocytes expressing the surface antigens CD4 and CD8. IFN gamma synthesis is induced by IL-2, FGF-basic, and EGF.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité