Human recombinant IL12
Fournisseur: ProSci Inc.
92-363
PRSI92-363EA 852 EUR InStock
PRSI92-363
Human recombinant IL12
Protéines et Peptides
IL-12 is involved in the differentiation of naive T cells into Th1 cells. It is known as a T cell-stimulating factor, which can stimulate the growth and function of T cells. It stimulates the production of interferon-gamma (IFN- gamma ) and tumour necrosis factor-alpha (TNF- alpha) from T cells and natural killer (NK) cells, and reduces IL-4 mediated suppression of IFN- gamma . T cells that produce IL-12 have a coreceptor, CD30, which is associated with IL-12 activity.IL-12 plays an important role in the activities of natural killer cells and T lymphocytes. IL-12 mediates enhancement of the cytotoxic activity of NK cells and CD8+ cytotoxic T lymphocytes.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité