Human recombinant EGFR (from cells)
Fournisseur: ProSci Inc.
92-366
PRSI92-366EA 852 EUR InStock
PRSI92-366
Human recombinant EGFR (from cells)
Protéines et Peptides
The EGFR subfamily of receptor tyrosine kinases is composed of EGFR, ErbB2, ErbB3 and ErbB4. The EGFR shares 43% - 44% aa sequence identity with the ECD of human EGFR subfamily. All these family members are type I transmembrane glycoproteins with an extracellular ligand binding domain. The extracellular ligand binding domain is containing two cysteine-rich domains separated by a spacer region and a cytoplasmic domain containing a membrane-proximal tyrosine kinase domain. Ligand binding could induce EGFR homodimerisation and heterodimerisation with ErbB2, resulting in cell signalling, heterodimerisation tyrosine phosphorylation and kinase activation. It can bind EGF, amphiregulin, TGF-alpha, betacellulin, epiregulin, HB-EGF, epigen, and so on. Its signalling regulates multiple biological functions including cell proliferation, differentiation, motility, and apoptosis. EGFR can also be recruited to form heterodimers with the ligand-activated ErbB3 or ErbB4. EGFR is overexpressed in different tumours. Several anti-cancer drugs use EGFR as target.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité