Mouse recombinant SEMA4A
Fournisseur: ProSci Inc.
92-371
PRSI92-371EA 852 EUR InStock
PRSI92-371
Mouse recombinant SEMA4A
Protéines et Peptides
Semaphorin-4A (SEMA4A) belongs to the semaphorin family which contains a Ig-like C2-type domain, a PSI domain and a Sema domain. SEMA4A is expressed from day 10 in the embryo, and low levels are found between days 10-12. SEMA4A is a cell surface receptor for PLXNB1, PLXNB2, PLXNB3 and PLXND1 that plays an important role in cell-cell signalling. It plays a role in priming antigen-specific T-cells, promotes differentiation of Th1 T-helper cells, and thereby contributes to adaptive immunity. SEMA4A promotes phosphorylation of TIMD2, inhibits angiogenesis, and promotes axon growth cone collapse, Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité