Mouse recombinant CD28
Fournisseur: ProSci Inc.
92-372
PRSI92-372EA 343 EUR InStock
PRSI92-372
Mouse recombinant CD28
Protéines et Peptides
T-cell-specific surface glycoprotein CD28, contains an Ig-like V-type domain. CD28 is one of the proteins expressed on T cells that provide co-stimulatory signals, which are required for their activation. It is the receptor for CD80 and CD86. When activated by Toll-like receptor ligands, the CD80 expression is upregulated in antigen presenting cells (APCs). The CD86 expression on antigen presenting cells is constitutive. CD28 is the only B7 receptor constitutively expressed on naive T cells. It involved in T-cell activation, the induction of cell proliferation and cytokine production and promotion of T-cell survival.
Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité