Human Recombinant Hyaluronan-binding 2 (from Cells)
Fournisseur: ProSci Inc.
92-373
PRSI92-373EA 852 EUR InStock
PRSI92-373
Human Recombinant Hyaluronan-binding 2 (from Cells)
Protéines et Peptides
Hyaluronan-binding protein 2(HABP2) is an extracellular serine protease which binds hyaluronic acid. It secreted as an inactive single-chain precursor and is then activated to a heterodimeric form, which consists of a 50 kDa heavy and a 27 kDa light chain linked by a disulfide bond. HABP2 is involved in cell adhesion, it can cleave the alpha-chain at multiple sites and the beta-chain between 'Lys-53' and 'Lys-54' , but not the gamma-chain of fibrinogen. As a result of this, it does not initiate the formation of the fibrin clot and does not cause the fibrinolysis directly. It does not cleave prothrombin and plasminogen but converts the inactive single chain urinary plasminogen activator to the active two chain form, activates coagulation factor VII.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité