Human recombinant MMP9 (from cells)
Fournisseur: ProSci Inc.
92-376
PRSI92-376EA 884 EUR InStock
PRSI92-376
Human recombinant MMP9 (from cells)
Protéines et Peptides
Matrix metallopeptidase 9 (MMP-9) is an enzyme encoded by the MMP9 gene. This protein, which is produced by normal alveolar macrophages and granulocytes, can be activated by 4-aminophenylmercuric acetate and phorbol ester and up-regulated by ARHGEF4, SPATA13 and APC via the JNK signalling pathway in colorectal tumour cells. MMP-9 is involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, angiogenesis, bone development, wound healling, cell migration, learning and memory, as well as in pathological processes, such as arthritis, intracerebral hemorrhage, and metastasis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité