Human recombinant D-tyrosyl-tRNA(Tyr) deacylase 1 (from cells)
Fournisseur: ProSci Inc.
92-377
PRSI92-377EA 884 EUR InStock
PRSI92-377
Human recombinant D-tyrosyl-tRNA(Tyr) deacylase 1 (from cells)
Enzymes
D-tyrosyl-tRNA(Tyr) deacylase 1(DTD1) belongs to the DTD family, and expressed in many adult and fetal tissues such as testis, ovary, spleen in adult and fetal brain. It is a nucleus and cytoplasm located protein, and is preferentially phosphorylated in cells arrested early in S phase. DTD1 is an ATPase involved in DNA replication, it may facilitate loading of CDC45 onto pre-replication complexes. The protein may hydrolyse D-tyrosyl-tRNA(Tyr) into D-tyrosine and free tRNA(Tyr), a possible defense mechanism against a harmful effect of D-tyrosine.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité