Human recombinant Complement factor H (from cells)
Fournisseur: ProSci Inc.
92-382
PRSI92-382EA 884 EUR InStock
PRSI92-382
Human recombinant Complement factor H (from cells)
Protéines et Peptides
Complement Factor H (CFH) is a secreted protein which is a member of the regulators of complement activation family and is a complement control protein. It is expressed by the liver and secreted in plasma. Its principal function is to regulate the Alternative Pathway of the complement system, ensuring that the complement system is directed towards pathogens or other dangerous material and does not damage host tissue. Factor H regulates complement activation on self cells and surfaces by possessing both cofactor activity for the Factor I mediated C3b cleavage, and decay accelerating activity against the alternative pathway C3-convertase, C3bBb. Factor H exerts its protective action on self cells and self surfaces but not on the surfaces of bacteria or viruses, because it binds to glycosaminoglycans (GAGs) that are generally present on host cells but not, normally, on pathogen surfaces.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité