Human recombinant Methylmalonyl-CoA epimerase (from cells)
Fournisseur: ProSci Inc.
92-387
PRSI92-387EA 409 EUR InStock
PRSI92-387
Human recombinant Methylmalonyl-CoA epimerase (from cells)
Enzymes
Methylmalonyl-CoA epimerase, mitochondrial(MCEE)is an enzyme which belongs to the glyoxalase I family. It converts (S)-methylmalonyl-CoA to the (R) form, catalyses the following chemical reaction: (R)-methylmalonyl-CoA (S)-methylmalonyl-CoA. It plays an important role in the catabolism of fatty acids with odd-length carbon chains. This protein deficiency is an autosomal recessive inborn error of AA metabolism, involving valine, threonine, isoleucine and methionine. This organic aciduria can appear in the neonatal period with life-threatening metabolic acidosis, hyperammonemia, feeding difficulties, pancytopenia and coma.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité