Human recombinant myelin P0-like 1 (from cells)
Fournisseur: ProSci Inc.
92-390
PRSI92-390EA 343 EUR InStock
PRSI92-390
Human recombinant myelin P0-like 1 (from cells)
Protéines et Peptides
Myelin protein zero-like protein 1(MPZL1) is encoded by the MPZL1 gene, which is a single-pass type I membrane protein. It is widely expressed with highest levels in heart, placenta, kidney and pancreas. As cell surface receptor, it involved in signal transduction processes. MPZL1 recruits PTPN11/SHP-2 to the cell membrane and subsequently activate/phosphorylate Src kinase at Tyr426, promoting phosphorylation of cortactin and migration of HCC cells. MPZL1also is a major receptor for concanavalin-A (ConA) and involved in cellular signalling induced by ConA, which probably includes Src family tyrosine-protein kinases.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité