Human Recombinant Ganglioside GM2 activator (from Cells)
Fournisseur: ProSci Inc.
92-391
PRSI92-391EA 761 EUR InStock
PRSI92-391
Human Recombinant Ganglioside GM2 activator (from Cells)
Protéines et Peptides
Ganglioside GM2 activator (GM2A) is a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta -hexosaminidase A (HEXB). HEXB together with GM2A, catalyses the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. GM2A accommodate several single chain phospholipids and fatty acids, is a lipid transfer protein that stimulates the enzymatic processing of gangliosides, and also T-cell activation through lipid presentation. It extracts single GM2 molecules from membranes and presents them in soluble form to beta-hexosaminidase A for cleavage of N-acetyl-D-galactosamine and conversion to GM3.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité