Human recombinant Selenophosphate synthase 1 (from cells)
Fournisseur: ProSci Inc.
92-396
PRSI92-396EA 789 EUR InStock
PRSI92-396
Human recombinant Selenophosphate synthase 1 (from cells)
Enzymes
Selenophosphate synthetase 1 (SEPHS1) belongs to the selenophosphate synthase 1 family, Class II subfamily. It has four different isoforms by alternative splicing. Isoform 1 and isoform 2 are gradually expressed during the cell cycle until G2/M phase and then decreased, which Isoform 3 is gradually expressed during the cell cycle until S phase and then decreased. SEPHS1 can be activated by phosphate ions and by potassium ions. It can synthesise synthesises selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesise selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité