Human recombinant MER (from cells)
Fournisseur: ProSci Inc.
92-397
PRSI92-397EA 761 EUR InStock
PRSI92-397
Human recombinant MER (from cells)
Protéines et Peptides
Tyrosine-protein kinase Mer (MERTK) is a single-pass type I membrane protein which belongs to the MER/AXL/TYRO3 receptor kinase family. MERTK include two fibronectin type-III domains, two Ig-like C2-type domains, and one tyrosine kinase domain. It can’t be expressed in normal B- and T-lymphocytes, but it is usually expressed in numerous neoplastic B- and T-cell lines. MERTK could regulate many physiological processes, such as cell survival, migration, differentiation. It was demonstrated that the MERTK plays critical role in the engulfment and efficient clearance of apoptotic cells, platelet aggregation, and cytoskeleton reorganisation. Not only these, it also plays an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signalling SOCS1 and SOCS3. In addition, MERTK could regulate rod outer segments fragments phagocytosis in the retinal pigment epithelium (RPE), deficiency in MERTK are the cause of retinitis pigmentosa.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité