Human Recombinant Coagulation factor IX (from Cells)
Fournisseur: ProSci Inc.
92-398
PRSI92-398EA 588 EUR InStock
PRSI92-398
Human Recombinant Coagulation factor IX (from Cells)
Protéines et Peptides
Coagulation factor IX(F9), is a member of the peptidase S1 family. It contains two EGF-like domains, a Gla domain and a peptidase S1 domain. It is primarily expressed in the liver and secreted in plasma. Factor IX is a vitamin K-dependent plasma protein that participates in the intrinsic pathway of blood coagulation by converting factor X to its active form in the presence of Ca2+ ions, phospholipids, and factor VIIIa. Mutations in position 43 and 46 prevents cleavage of the propeptide, mutation in position 93 probably fails to bind to cell membranes, mutation in position 191 or in position 226 prevent cleavage of the activation peptide. Mutations of human F9 can result in thrombophilia and recessive X-linked hemophilia B (HEMB). An X-linked blood coagulation disorder characterized by a permanent tendency to hemorrhage, due to factor IX deficiency. It is phenotypically similar to hemophilia A, but patients present with fewer symptoms. Many patients are asymptomatic until the hemostatic system is stressed by surgery or trauma.
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité