Human recombinant serpin B12 (from cells)
Fournisseur: ProSci Inc.
92-399
PRSI92-399EA 761 EUR InStock
PRSI92-399
Human recombinant serpin B12 (from cells)
Protéines et Peptides
Serpin B12 is a member of the serpin family. Serpins are the largest and most diverse family of serine protease inhibitors. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. Serpin B12 is expressed in many tissues, including brain, bone marrow, lymph node, heart, lung, liver, pancreas, testis, ovary, and intestine. Serpins are involved in a number of fundamental biological processes such as blood coagulation, complement activation, fibrinolysis, angiogenesis, inflammation and tumour suppression and are expressed in a cell-specific manner. SerpinB12 inhibits trypsin and plasmin, but not thrombin, coagulation factor Xa, or urokinase-type plasminogen activator.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité