Mouse recombinant TRAILR2
Fournisseur: ProSci Inc.
92-400
PRSI92-400EA 588 EUR InStock
PRSI92-400
Mouse recombinant TRAILR2
Protéines et Peptides
Mouse tumour necrosis factor receptor superfamily member 10B (TNFRSF10B) is a member of the TNFR family which contains 1 death domain and 3 TNFR-Cys repeats. TNFRSF10B exhibits high structural and functional homology to TRAIL-R1 (DR-4). TNFRSF10B is highly expressed in heart, lung, lymphocytes, spleen and kidney. In addition, it is regulated by the tumour suppressor p53. TNFRSF10B is the receptor for the cytotoxic ligand TNFSF10/TRAIL. It promotes the activation of NF-kappa-B and is essential for ER stress-induced apoptosis. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signalling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases mediating apoptosis.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité