Human recombinant Cryptic (from cells)
Fournisseur: ProSci Inc.
92-402
PRSI92-402EA 588 EUR InStock
PRSI92-402
Human recombinant Cryptic (from cells)
Protéines et Peptides
Cryptic (CFC1) is a member of the epidermal growth factor (EGF)- Cripto, Frl-1, and Cryptic (CFC) family. It contains an EGF-like domain, and is glycosylated on its N-terminal during post-translational modification. Cryptic is identified as a NODAL coreceptor involved in the correct establishment of the left-right axis. It may play a role in mesoderm and/or neural patterning during gastrulation. The unnormal expression of this gene may causes a series of diseases such as HTX2, Transposition of the great arteries dextro-looped 2, and Conotruncal heart malformations.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité