Human recombinant serpin I2 (from cells)
Fournisseur: ProSci Inc.
92-404
PRSI92-404EA 654 EUR InStock
PRSI92-404
Human recombinant serpin I2 (from cells)
Protéines et Peptides
SERPINI2 is a secreted protein of the serpin family. Serpins are a group of proteins with similar structures that were first identified as a set of proteins able to inhibit proteases. As protease inhibitors, serpins have an array of functions including regulating blood coagulation, fibrinolysis, the complement pathway, angiogenesis, inflammation, tumour suppression, extracellular matrix remodelling, and cell motility. SERPINI2 is expressed in human tissues including pancreas and adipose tissues. Mutations of human SERPINI2 can directly result in conditions such as acinar cell apoptosis and malabsorption.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité