Mouse Recombinant Coagulation factor X
Fournisseur: ProSci Inc.
92-406
PRSI92-406EA 852 EUR InStock
PRSI92-406
Mouse Recombinant Coagulation factor X
Protéines et Peptides
Mouse coagulation factor X / F10 a member of the peptidase S1 family. The mature F10 is composed mostly of two EGF-like domains, one Gla gamma-carboxy-glutamate domain and one peptidase S1 domain. Factor Xa is a vitamin K-dependent plasma protease that converts prothrombin to thrombin in the presence of factor Va, calcium and phospholipid during blood clotting. The two chains of F10 are formed from a single-chain precursor by the excision of two Arg residues. A single-chain precursor is initially Synthesised in the liver. The light and heavy chains are linked together by disulfide bonds. The light chain contains a Gla and two EGF-like domains. The heavy chain corresponds to the serine protease domain. It can form a heterodimer with SERPINA5.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité