Mouse recombinant Apolipoprotein E
Fournisseur: ProSci Inc.
92-407
PRSI92-407EA 813 EUR InStock
PRSI92-407
Mouse recombinant Apolipoprotein E
Protéines et Peptides
Apolipoprotein E (Apo-E), is a member of the apolipoprotein A1/A4/E family. APOE may function in mediating the binding, internalisation, and catabolism of lipoprotein particles. It can serve as a ligand for the LDL (apo B/E) receptor and for the specific apo-E receptor (chylomicron remnant) of hepatic tissues. APOE is usually secreted in plasma. Phosphorylation sites are present in the extracellular medium.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité