Mouse recombinant nectin-2
Fournisseur: ProSci Inc.
92-408
PRSI92-408EA 409 EUR InStock
PRSI92-408
Mouse recombinant nectin-2
Protéines et Peptides
Nectin-2( CD112) is a member of the nectin family, which contains two Ig-like C2-type domains and one Ig-like V-type domain in the extracellular region. Nectins are type I transmembrane glycoproteins that are calcium-independent immunoglobulin (Ig)-like cell adhesion molecules (CAMs). Nectin2 is widely expressed in human tissues including brain, spinal cord, spleen, kidney, heart and liver. It can form trans-heterodimers with PVRL3/nectin-3 and interacts with CD226. Mutations of alleles of the murine CD112 gene can result in conditions such as morphologically aberrant spermatozoa. It may function in allergic reactions, and accordingly may used as a novel target for anti-allergic therapy.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité