Mouse recombinant Siglec-3
Fournisseur: ProSci Inc.
92-412
PRSI92-412EA 654 EUR InStock
PRSI92-412
Mouse recombinant Siglec-3
Protéines et Peptides
Mouse myeloid cell surface antigen CD33(CD33) is a member of the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. CD33 contains one Ig-like C2-type domain and one Ig-like V-type domain. CD33 is a putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. CD33 preferentially binds to alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, CD33 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signalling molecules. CD33 induces apoptosis in acute myeloid leukemia. CD33 is becoming increasingly important as a target of antibody-mediated therapy in acute myeloid leukaemia (AML).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité