Mouse recombinant Syndecan 4
Fournisseur: ProSci Inc.
92-413
PRSI92-413EA 365 EUR InStock
PRSI92-413
Mouse recombinant Syndecan 4
Protéines et Peptides
Mouse SDC4 is a ubiquitous transmembrane proteoglycan which belongs to the syndecan proteoglycan family. SDC4 is a cell surface proteoglycan that bears heparan sulphate. The four vertebrate syndecans, Syndecan-1 through -4, have similar short cytoplasmic domains and extracellular portions that diverge, except for HS attachment sites. Structurally diverse side chains add considerably to the size of the core proteins and serve as binding sites for growth factors, cytokines, and extracellular matrix proteins. Syndecans are present as homodimers or multimers, and are often expressed in developmental and cell type-specific patterns. It is expressed highly in liver, kidney and lung. SDC4 localises to the focal adhesions of adherent cells and binds to a range of extracellular ligands, including growth factors and extracellular-matrix proteins. Through its extracellular domain, syndecan-4 cooperates with adhesion molecules and binds matrix components relevant for cell migration. As a heparan sulphate proteoglycan, SDC4 works as a coreceptor for various growth factors. Syn4 deficiency limits neointimal formation after vascular injury by regulating vascular smooth muscle cells (VSMCs) proliferation and vascular progenitor cells (VPCs) mobilisation. SDC4 have an array of functions including regulating cell growth, differentiation, and adhesion.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité