Mouse recombinant carboxypeptidase A4
Fournisseur: ProSci Inc.
92-415
PRSI92-415EA 884 EUR InStock
PRSI92-415
Mouse recombinant carboxypeptidase A4
Enzymes
Carboxypeptidase A4 (CPA4) is a member of the peptidase M14 family. CPA4 is metalloprotease that could be involved in the histone hyperacetylation pathway. CPA4 binds one zinc ion per subunit and could catalyse to release of a C-terminal amino acid, with preference for -Phe, -Leu, -Ile, -Met, -Tyr and -Val.They have distinct expression patterns and different specificities for example, preferentially cleaving aromatic (carboxypeptidase As) or basic (carboxypeptidase Bs) residues. Several, such as carboxypeptidase Xs, have lost their catalytic activity. Carboxypeptidases play important roles in digestion of food, processing of bioactive peptides and blood coagulation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité