Mouse recombinant carboxypeptidase A2
Fournisseur: ProSci Inc.
92-419
PRSI92-419EA 654 EUR InStock
PRSI92-419
Mouse recombinant carboxypeptidase A2
Enzymes
Mouse carboxypeptidase A2(CPA2) is a secreted pancreatic procarboxy -peptidase which belongs to the peptidase M14 family. CPA2 consists of a signal peptide, a pro region and a mature chain. It can be activated after cleavage of the pro peptide. CPA2 cleaves the C-terminal amide or ester bond of peptides that have a free C-terminal carboxyl group. The hydrolytic action of CPA2 was identified with a preference towards long substrates with aromatic amino acids in their C-terminal end, particularly tryptophan.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité