Human recombinant Chemerin (from cells)
Fournisseur: ProSci Inc.
92-423
PRSI92-423EA 584 EUR InStock
PRSI92-423
Human recombinant Chemerin (from cells)
Protéines et Peptides
Retinoic acid receptor responder protein 2(RARRES2) is a secreted protein that in humans is encoded by the RARRES2 gene. It is highly expressed in skin, also found in pancreas, liver, spleen, prostate, ovary, small intestine and colon. It is a chemoattractant protein that acts as a ligand for the G protein-coupled receptor CMKLR1. RARRES2 is secreted in an inactive form as prochemerin and is activated through cleavage of the C-terminus by inflammatory and coagulation serine proteases. It is thought to act as a cell surface receptor, found to stimulate chemotaxis of dendritic cells and macrophages to the site of inflammation. RARRES2 is inhibited in psoriatic lesions,it is activated by tazarotene in skin rafts and in the epidermis of psoriatic lesions.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité