Mouse recombinant Ephrin B1
Fournisseur: ProSci Inc.
92-425
PRSI92-425EA 306 EUR InStock
PRSI92-425
Mouse recombinant Ephrin B1
Protéines et Peptides
Mouse Ephrin-B1 is a single-pass type I membrane protein which belongs to the ephrin family. It contains an ephrin RBD (ephrin receptor-binding) domain, and expressed in heart, placenta, lung, liver, skeletal muscle, kidney and pancreas. Ephrin-B1 is cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. It binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signalling into neighboring cells. It may play a role in cell adhesion and function in the development or maintenance of the nervous system.
Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité