Human recombinant IL12 p40 (from cells)
Fournisseur: ProSci Inc.
92-427
PRSI92-427EA 654 EUR InStock
PRSI92-427
Human recombinant IL12 p40 (from cells)
Protéines et Peptides
Interleukin-12 subunit beta (IL-12B) belongs to the type I cytokine receptor family. It contains 1 fibronectin type-III domain and 1 Ig-like C2-type domain. IL-12B is a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. IL-12 is a disulfide-linked heterodimer composed of the 40 kD cytokine receptor encoded by IL12B and a 35 kD subunit encoded by IL12A. IL12 is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. It has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité