Human recombinant CXCL4 (from cells)
Fournisseur: ProSci Inc.
92-429
PRSI92-429EA 584 EUR InStock
PRSI92-429
Human recombinant CXCL4 (from cells)
Protéines et Peptides
Human Chemokine (C-X-C motif) Ligand 4 (CXCL4) is expressed in megakaryocytes and stored in the alpha-granules of platelets. CXCL4 contains several heparin-binding sites at the C-terminal region and binds heparin with high affinity. The active CXCL4 protein is a tetramer. Human and mouse CXCL4 share 64% sequence identity. CXCL4 is chemotactic for neutrophils, fibroblasts and monocytes and plays a critical role in inflammation and wound repair. CXCL4 functions via a splice variant of the chemokine receptor CXCR3, known as CXCR3B. The major physiologic role of CXCL4 appears to be neutralisation of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. In contrast to other CXC chemokines, CXCL4 lacks chemotactic activity for polymorphonuclear granulocytes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité