Human Recombinant GDNF Receptor alpha 2 (from Cells)
Fournisseur: ProSci Inc.
92-432
PRSI92-432EA 306 EUR InStock
PRSI92-432
Human Recombinant GDNF Receptor alpha 2 (from Cells)
Protéines et Peptides
GDNF family receptor alpha-2 is a glycosylphosphatidylinosito l (GPI)-linked cell surface receptor. It is part of the GDNF receptor family. Glial cell line-derived neurotrophic factor (GDNF) and neurturin (NTN) are two structurally related, potent neurotrophic factors that play key roles in the control of neuron survival and differentiation. GFRA2 mediates the NRTN-induced autophosphorylation and activation of the RET receptor. It also able to mediate GDNF signalling through the RET tyrosine kinase receptor. It acts preferentially as a receptor for NTN compared to its other family member, GDNF family receptor alpha 1.
Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité