Human recombinant serpin B9 (from cells)
Fournisseur: ProSci Inc.
92-434
PRSI92-434EA 588 EUR InStock
PRSI92-434
Human recombinant serpin B9 (from cells)
Protéines et Peptides
Serpin B9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. Serpin B9 is an inhibitor of the granzyme B/perforin lytic pathway. It is expressed in normal mammary epithelial cells but not in most mammary carcinoma cell lines. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodelling, and apoptosis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité