Human recombinant Cytochrome b5 B (from cells)
Fournisseur: ProSci Inc.
92-437
PRSI92-437EA 761 EUR InStock
PRSI92-437
Human recombinant Cytochrome b5 B (from cells)
Protéines et Peptides
Cytochrome b5 type B (CYB5B) is a membrane of the cytochrome b5 family. It contains 1 cytochrome b5 heme-binding domain. Cytochrome b5 is a membrane bound hemoprotein which function as an electron carrier for several membrane bound oxygenases. In the mitochondrion of eukaryotes and in aerobic prokaryotes, cytochrome b is a component of respiratory chain complex III also known as the bc1 complex or ubiquinol-cytochrome c reductase.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité