Human recombinant periostin (from cells)
Fournisseur: ProSci Inc.
92-440
PRSI92-440EA 525 EUR InStock
PRSI92-440
Human recombinant periostin (from cells)
Protéines et Peptides
Periostin is a secreted, homodimeric protein that belongs to the periostin family of the FAS1 superfamily of molecules. It is a TGF-beta inducible molecule that serves as both an adhesion molecule and tumour suppressor. It is synthesised by smooth muscle cells, fibroblasts and osteoblasts, as well as in the periosteum and periodontal ligament.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité