Human recombinant transferrin (from cells)
Fournisseur: ProSci Inc.
92-442
PRSI92-442EA 409 EUR InStock
PRSI92-442
Human recombinant transferrin (from cells)
Protéines et Peptides
Serotransferrin belongs to transferrin family, and contains 2 transferrin-like domains. The protein is a secreted protein, and expressed by the liver and secreted in plasma. Transferrins are iron binding transport proteins which can bind two Fe3+ ions in association with the binding of an anion. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilisation. Serum transferrin may also have a further role in stimulating cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité