Human recombinant OX40 Ligand (from cells)
Fournisseur: ProSci Inc.
92-445
PRSI92-445EA 852 EUR InStock
PRSI92-445
Human recombinant OX40 Ligand (from cells)
Protéines et Peptides
tumour necrosis factor ligand superfamily member 4(TNFSF4/OX40L) is a single-pass type II membrane protein. OX40L is the ligand for CD134 and is expressed on such cells as DC2s enabling amplification of Th2 cell differentiation. It is a cytokine that binds to TNFRSF4, Co-stimulates T-cell proliferation and cytokine production. It has been found association with systemic lupus erythematosus, No association with occurrence of atherosclerosis.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité