Mouse recombinant GMCSF
Fournisseur: ProSci Inc.
92-446
PRSI92-446EA 761 EUR InStock
PRSI92-446
Mouse recombinant GMCSF
Protéines et Peptides
Granulocyte-Macrophage Colony Stimulating Factor (GM-CSF) was initially characterised as a growth factorthat can support the in vitro colony formation of granulocyte-macrophage progenitors. It is produced by anumber of different cell types (including activated T cells, B cells, macrophages, mast cells, endothelial cellsand fibroblasts) in response to cytokine of immune and inflammatory stimuli. Besides granulocyte-macrophageprogenitors, GM-CSF is also a growth factor for erythroid, megakaryocyte and eosinophil progenitors. Onmature hematopoietic, monocytes/ macrophages and eosinophils. GM-CSF has a functional role on nonhematopoitic cells. It can induce human endothelial cells to migrate and proliferate. Additionally, GM-CSF canalso stimulate the proliferation of a number of tumour cell lines, including osteogenic sarcoma, carcinoma andadenocarcinoma cell lines.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité