Mouse recombinant HLADG
Fournisseur: ProSci Inc.
92-450
PRSI92-450EA 525 EUR InStock
PRSI92-450
Mouse recombinant HLADG
Protéines et Peptides
Mouse HLA class II histocompatibility antigen gamma chain (CD74), is a single-pass type II membrane protein that in humans is encoded by the CD74 gene. It contains 1 thyroglobulin type-1 domain. CD74 Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to compartments where peptide loading of class II takes place.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-mFc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité