Human recombinant TIMP-4 (from cells)
Fournisseur: ProSci Inc.
92-458
PRSI92-458EA 852 EUR InStock
PRSI92-458
Human recombinant TIMP-4 (from cells)
Protéines et Peptides
Metalloproteinase inhibitor 4 is an enzyme that in humans is encoded by the TIMP4 gene, belongs to the protease inhibitor I35 (TIMP) family. The protein complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Known to act on MMP-1, MMP-2, MMP-3, MMP-7 and MMP-9.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité