Mouse recombinant cathepsin B
Fournisseur: ProSci Inc.
92-463
PRSI92-463EA 852 EUR InStock
PRSI92-463
Mouse recombinant cathepsin B
Protéines et Peptides
Cathepsin B (CatB) is an enzymatic protein belonging to the peptidase (or protease) families. which is believed to participate in intracellular degradation and turnover of proteins. Has also been implicated in tumour invasion and metastasis。
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité